missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, 99.9%, Honeywell™

Catalog No. p-7120024
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047079 1 L Stainless Steel Can
60047080 2.5 L Glass Bottle
60047084 6 x 1 L Glass Bottle
60047083 4 x 2.5 L Glass Bottle
60047081 20 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60047079 Supplier Honeywell Chemicals Supplier No. 5379931L
Only null left

≥99.9%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point ≤-90°C
Appearance Liquid
Density 0.81g/mL at 25°C
Boiling Point 119°C
Quantity 1 L
Explosion Limit 11.2%
Ignition Point 649°C
Packaging Stainless Steel Can
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Molecular Weight (g/mol) 74.123
Formula Weight 74.12g/mol
Percent Purity 99%
Chemical Name or Material 1-Butanol
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
DOTInformation class 3 - PG 3 - Butanols
RTECSNumber 200-751-6