missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol 97.0+%, TCI America™

Supplier:  TCI America B0685500ML

 View more versions of this product

Catalog No. B0685500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Butanethiol
Colorless
C4H10S
500 mL
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
CCCCS
90.184
90.18
Liquid
109-79-5
98°C
MFCD00004905
2347
WQAQPCDUOCURKW-UHFFFAOYSA-N
butane-1-thiol
8012
≥97.0% (GC)

Chemical Identifiers

109-79-5
90.184
WQAQPCDUOCURKW-UHFFFAOYSA-N
8012
CCCCS
C4H10S
MFCD00004905
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
butane-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0464

RTECSNumber : EK6300000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions