missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol 97.0+%, TCI America™

Catalog No. p-7118611
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B068525ML 25 mL
B0685500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B068525ML Supplier TCI America Supplier No. B068525ML
Only null left

Chemical Identifiers

CAS 109-79-5
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.184
MDL Number MFCD00004905
InChI Key WQAQPCDUOCURKW-UHFFFAOYSA-N
Synonym 1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
PubChem CID 8012
IUPAC Name butane-1-thiol
SMILES CCCCS

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 mL
UN Number 2347
Formula Weight 90.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butanethiol
EINECSNumber (2)-0464
RTECSNumber EK6300000
TSCA Yes