missing translation for 'onlineSavingsMsg'
Learn More

1-Butanesulfonyl Chloride 98.0+%, TCI America™

Catalog No. B086625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B086625G 25 g
B0866500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B086625G Supplier TCI America Supplier No. B086625G

Chemical Identifiers

CAS 2386-60-9
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.624
MDL Number MFCD00007464
InChI Key WEDIIKBPDQQQJU-UHFFFAOYSA-N
Synonym 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
PubChem CID 75430
IUPAC Name butane-1-sulfonyl chloride
SMILES CCCCS(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 65°C
Quantity 25 g
UN Number 3265
Formula Weight 156.62
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butanesulfonyl Chloride
RTECSNumber EK5950000
TSCA Yes