Learn More
1-Butanesulfonyl chloride, 98%
CAS: 2386-60-9 | C4H9ClO2S | 156.624 g/mol
$55.08 - $474.86
Chemical Identifiers
| CAS | 2386-60-9 |
|---|---|
| Molecular Formula | C4H9ClO2S |
| Molecular Weight (g/mol) | 156.624 |
| MDL Number | MFCD00007464 |
| InChI Key | WEDIIKBPDQQQJU-UHFFFAOYSA-N |
| Synonym | 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride |
| PubChem CID | 75430 |
| IUPAC Name | butane-1-sulfonyl chloride |
| SMILES | CCCCS(=O)(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1829506
|
Thermo Scientific Chemicals
A1829506 |
5 g |
Each for $55.08
|
|
|||||
|
AAA1829522
|
Thermo Scientific Chemicals
A1829522 |
100 g |
Each for $474.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2386-60-9 | |
| 156.624 | |
| WEDIIKBPDQQQJU-UHFFFAOYSA-N | |
| 75430 | |
| CCCCS(=O)(=O)Cl |
| C4H9ClO2S | |
| MFCD00007464 | |
| 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride | |
| butane-1-sulfonyl chloride |
Specifications
| 2386-60-9 | |
| 1.21 | |
| 79°C (174°F) | |
| 1.456 | |
| MFCD00007464 | |
| UN3265 | |
| Moisture sensitive | |
| WEDIIKBPDQQQJU-UHFFFAOYSA-N | |
| butane-1-sulfonyl chloride | |
| 75430 | |
| 98% |
| -29°C | |
| 80°C to 81°C (9 mmHg) | |
| C4H9ClO2S | |
| CH3(CH2)3SO2Cl | |
| 5 g | |
| 1748742 | |
| 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride | |
| CCCCS(=O)(=O)Cl | |
| 156.624 | |
| 156.63 | |
| 1-Butanesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 219-204-8
RTECSNumber : EK5950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only