missing translation for 'onlineSavingsMsg'
Learn More

1-Butanesulfonyl chloride, 98%

Catalog No. AAA1829506
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1829506 5 g
AAA1829522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1829506 Supplier Thermo Scientific Chemicals Supplier No. A1829506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2386-60-9 | C4H9ClO2S | 156.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2386-60-9
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.624
MDL Number MFCD00007464
InChI Key WEDIIKBPDQQQJU-UHFFFAOYSA-N
Synonym 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
PubChem CID 75430
IUPAC Name butane-1-sulfonyl chloride
SMILES CCCCS(=O)(=O)Cl

Specifications

Melting Point -29°C
Density 1.21
Boiling Point 80°C to 81°C (9 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.456
Linear Formula CH3(CH2)3SO2Cl
Quantity 5 g
UN Number UN3265
Beilstein 1748742
Sensitivity Moisture sensitive
Formula Weight 156.63
Percent Purity 98%
Chemical Name or Material 1-Butanesulfonyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 219-204-8
RTECSNumber EK5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only