missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoundecane 98.0+%, TCI America™
$74.56 - $559.22
Chemical Identifiers
| CAS | 693-67-4 |
|---|---|
| Molecular Formula | C11H23Br |
| Molecular Weight (g/mol) | 235.209 |
| MDL Number | MFCD00000223 |
| InChI Key | IKPSIIAXIDAQLG-UHFFFAOYSA-N |
| Synonym | undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane |
| PubChem CID | 12744 |
| IUPAC Name | 1-bromoundecane |
| SMILES | CCCCCCCCCCCBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B093425ML
|
TCI America
B093425ML |
25 mL |
Each for $74.56
|
|
|||||
|
B0934500ML
|
TCI America
B0934500ML |
500 mL |
Each for $559.22
|
|
|||||
Chemical Identifiers
| 693-67-4 | |
| 235.209 | |
| IKPSIIAXIDAQLG-UHFFFAOYSA-N | |
| 12744 | |
| CCCCCCCCCCCBr |
| C11H23Br | |
| MFCD00000223 | |
| undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane | |
| 1-bromoundecane |
Specifications
| 693-67-4 | |
| Yellow | |
| C11H23Br | |
| 25 mL | |
| IKPSIIAXIDAQLG-UHFFFAOYSA-N | |
| 1-bromoundecane | |
| 12744 | |
| ≥98.0% (GC) | |
| 1-Bromoundecane |
| -9°C | |
| 138°C | |
| MFCD00000223 | |
| undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane | |
| CCCCCCCCCCCBr | |
| 235.209 | |
| 235.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes