missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoundecane 98.0+%, TCI America™

Catalog No. B0934500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0934500ML 500 mL
B093425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0934500ML Supplier TCI America Supplier No. B0934500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 693-67-4
Molecular Formula C11H23Br
Molecular Weight (g/mol) 235.209
MDL Number MFCD00000223
InChI Key IKPSIIAXIDAQLG-UHFFFAOYSA-N
Synonym undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
PubChem CID 12744
IUPAC Name 1-bromoundecane
SMILES CCCCCCCCCCCBr

Specifications

Melting Point -9°C
Color Yellow
Boiling Point 138°C
Quantity 500 mL
Formula Weight 235.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromoundecane
EINECSNumber (2)-0067
TSCA Yes