missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoundecane, 98%

Catalog No. AAA1433336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1433336 500 g
AAA1433314 25 g
AAA1433322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1433336 Supplier Thermo Scientific Chemicals Supplier No. A1433336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 693-67-4 | C11H23Br | 235.209 g/mol

1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 693-67-4
Molecular Formula C11H23Br
Molecular Weight (g/mol) 235.209
MDL Number MFCD00000223
InChI Key IKPSIIAXIDAQLG-UHFFFAOYSA-N
Synonym undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
PubChem CID 12744
IUPAC Name 1-bromoundecane
SMILES CCCCCCCCCCCBr

Specifications

Melting Point -9°C
Density 1.052
Boiling Point 112°C to 114°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4563
Quantity 500 g
Beilstein 1098390
Solubility Information Insoluble in water.
Formula Weight 235.22
Percent Purity 98%
Chemical Name or Material 1-Bromoundecane
Show More Show Less
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-757-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only