missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoundecane, 98%

Catalog No. AC224010250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224010250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC224010250 Supplier Thermo Scientific Chemicals Supplier No. 224010250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 693-67-4 | C11H23Br | 235.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 693-67-4
MDL Number MFCD00000223
InChI Key IKPSIIAXIDAQLG-UHFFFAOYSA-N
Synonym undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
PubChem CID 12744
IUPAC Name 1-bromoundecane
SMILES CCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -9°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 137°C to 138°C (18.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C11H23Br
Refractive Index 1.4555 to 1.4575
Linear Formula CH3(CH2)10Br
Quantity 25 g
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 235.21
Formula Weight 235.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromoundecane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-757-3
TSCA TSCA

RUO – Research Use Only