missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane 97.0+%, TCI America™

Catalog No. B0233500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0233500ML 500 mL
B023325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0233500ML Supplier TCI America Supplier No. B0233500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.29
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 205°C
Quantity 500 mL
Formula Weight 277.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromotetradecane
EINECSNumber (2)-0067
TSCA Yes