missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane, 98%

Catalog No. p-77188
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187381000 100 g Glass bottle
AC187385000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC187381000 Supplier Thermo Scientific Chemicals Supplier No. 187381000
Only null left

CAS: 112-71-0 | C14H29Br | 277.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.28
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 175.0°C to 178.0°C (20.0 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4590 to 1.4610
Linear Formula CH3(CH2)13Br
Quantity 100 g
Beilstein 01,II,136
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 277.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromotetradecane, 98%
Show More Show Less
EINECSNumber 203-999-3
TSCA TSCA

RUO – Research Use Only