missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopyrene 95.0+%, TCI America™

Catalog No. p-7119082
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14955G 5 g
B149525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14955G Supplier TCI America Supplier No. B14955G
Only null left

Chemical Identifiers

CAS 1714-29-0
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.152
MDL Number MFCD00015767
InChI Key HYGLETVERPVXOS-UHFFFAOYSA-N
Synonym 1-bromo-pyrene, pyrene, bromo, pyrene, 1-bromo, bromopyrene, 1-brompyren, 1-brpy, 3-bromopyrene, 1-bromanylpyrene, pubchem14567, 1-bromopyrene
PubChem CID 159627
IUPAC Name 1-bromopyrene
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)Br

Specifications

Melting Point 98°C
Color White-Yellow
Boiling Point 255°C
Quantity 5 g
Formula Weight 281.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromopyrene