missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopyrene, 95%

Catalog No. p-7049682
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1828003 1 g
AA1828006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1828003 Supplier Thermo Scientific Chemicals Supplier No. 01828003
Only null left

CAS: 1714-29-0 | C16H9Br | 281.152 g/mol

1-Bromopyrene is used in the cleavage of DNA in studies. It is also used in the preparation of DNA base analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1714-29-0
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.152
MDL Number MFCD00015767
InChI Key HYGLETVERPVXOS-UHFFFAOYSA-N
Synonym 1-bromo-pyrene, pyrene, bromo, pyrene, 1-bromo, bromopyrene, 1-brompyren, 1-brpy, 3-bromopyrene, 1-bromanylpyrene, pubchem14567, 1-bromopyrene
PubChem CID 159627
IUPAC Name 1-bromopyrene
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)Br

Specifications

Melting Point 94°C to 95°C
Quantity 1 g
Formula Weight 281.13
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material 1-Bromopyrene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only