missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopropane 98.0+%, TCI America™

Catalog No. p-7118581
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B063825G 25 g
B0638500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B063825G Supplier TCI America Supplier No. B063825G
Only null left

Chemical Identifiers

CAS 106-94-5
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.993
MDL Number MFCD00000254
InChI Key CYNYIHKIEHGYOZ-UHFFFAOYSA-N
Synonym n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
PubChem CID 7840
ChEBI CHEBI:47105
IUPAC Name 1-bromopropane
SMILES CCCBr

Specifications

Melting Point -110°C
Color Colorless
Boiling Point 71°C
Quantity 25 g
UN Number 2344
Formula Weight 122.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromopropane
EINECSNumber (2)-0073
RTECSNumber TX4110000
TSCA Yes