Learn More
1-Bromopropane, 99%
CAS: 106-94-5 | C3H7Br | 122.993 g/mol
Supplier: Thermo Scientific Chemicals A104610J
Description
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.
Solubility
Miscible with ethanol, water and diethyl ether.
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| 1-Bromopropane | |
| -110°C | |
| 70°C to 71°C | |
| C3H7Br | |
| CH3CH2CH2Br | |
| 5000 mL | |
| 505936 | |
| 14,7845 | |
| Miscible with ethanol,water and diethyl ether. | |
| CCCBr | |
| 122.993 | |
| CHEBI:47105 | |
| 99% |
| 106-94-5 | |
| 1.353 | |
| 21°C (70°F) | |
| 1.434 | |
| MFCD00000254 | |
| UN2344 | |
| Light sensitive | |
| n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide | |
| CYNYIHKIEHGYOZ-UHFFFAOYSA-N | |
| 1-bromopropane | |
| 7840 | |
| 123 |
Chemical Identifiers
| 106-94-5 | |
| 122.993 | |
| CYNYIHKIEHGYOZ-UHFFFAOYSA-N | |
| 7840 | |
| 1-bromopropane |
| C3H7Br | |
| MFCD00000254 | |
| n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide | |
| CHEBI:47105 | |
| CCCBr |
Safety and Handling
GHS H Statement
H225-H360-H373-H315-H319-H335-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335-H336-H350-H360FD-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOPROPANES
EINECSNumber : 203-445-0
RTECSNumber : TX4110000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only