missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopropane, 98%, Spectrum™ Chemical

Catalog No. 18608086
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
250 kg
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608086 250 mL Amber Glass Bottle
18608083 1 L Amber Glass Bottle
18608087 4 L Amber Glass Bottle
18608084 20 L Steel Can
18608085 250 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608086 Supplier Spectrum Chemical Mfg Cor Supplier No. B1157250ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 106-94-5

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 250 mL
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.