missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopropane, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 71°C/159.8°F |
| Packaging | Amber Glass Bottle |
| Quantity | 20 L |
| Molecular Weight (g/mol) | 123.00 |
| Percent Purity | 0.98 |
| Grade | Reagent |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Bromopropane |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.