missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentane, 98%

Catalog No. p-76348
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152522500 250 mL Glass bottle
AC152520010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152522500 Supplier Thermo Scientific Chemicals Supplier No. 152522500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-53-2 | C5H11Br | 151.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-53-2
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000267
InChI Key YZWKKMVJZFACSU-UHFFFAOYSA-N
Synonym n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0
PubChem CID 8057
IUPAC Name 1-bromopentane
SMILES CCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -95°C
Color Colorless to Yellow
Density 1.2100g/mL
Boiling Point 130°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.446
Linear Formula CH3(CH2)4Br
Quantity 250 mL
Beilstein 01, 131
Specific Gravity 1.21
Merck Index 15, 596
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,miscible with ether
Formula Weight 151.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromopentane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-776-0
RTECSNumber RZ9770000
TSCA TSCA

RUO – Research Use Only