missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctadecane 97.0+%, TCI America™

Catalog No. p-7118571
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B062525G 25 g
B0625500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B062525G Supplier TCI America Supplier No. B062525G
Only null left

Chemical Identifiers

CAS 112-89-0
Molecular Formula C18H37Br
Molecular Weight (g/mol) 333.40
MDL Number MFCD00000231
InChI Key WSULSMOGMLRGKU-UHFFFAOYSA-N
Synonym octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
PubChem CID 8218
IUPAC Name 1-bromooctadecane
SMILES CCCCCCCCCCCCCCCCCCBr

Specifications

Quantity 25 g
Formula Weight 333.40
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromooctadecane
EINECSNumber (2)-0067
TSCA Yes