missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctadecane, 97%

Catalog No. p-7022696
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1558822 100 g
AAA1558836 500 g
AAA155880E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1558822 Supplier Thermo Scientific Chemicals Supplier No. A1558822

CAS: 112-89-0 | C18H37Br | 333.40 g/mol

1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.

Solubility
Soluble in acetone and most common organic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 112-89-0
Molecular Formula C18H37Br
Molecular Weight (g/mol) 333.40
MDL Number MFCD00000231
InChI Key WSULSMOGMLRGKU-UHFFFAOYSA-N
Synonym octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
PubChem CID 8218
IUPAC Name 1-bromooctadecane
SMILES CCCCCCCCCCCCCCCCCCBr

Specifications

Melting Point 25°C to 30°C
Density 0.976
Boiling Point 150°C to 152°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Linear Formula CH3(CH2)17Br
Quantity 100 g
Beilstein 774145
Solubility Information Soluble in acetone and most common organic solvents. Insoluble in water.
Formula Weight 333.39
Percent Purity 97%
Chemical Name or Material 1-Bromooctadecane
Show More Show Less
EINECSNumber 204-013-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only