missing translation for 'onlineSavingsMsg'
Learn More

1-Bromononane 98.0+%, TCI America™

Catalog No. p-7118753
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B093325ML 25 mL
B0933100ML 100 mL
B0933500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B093325ML Supplier TCI America Supplier No. B093325ML
Only null left

Chemical Identifiers

CAS 693-58-3
Molecular Formula C9H19Br
Molecular Weight (g/mol) 207.155
MDL Number MFCD00000278
InChI Key AYMUQTNXKPEMLM-UHFFFAOYSA-N
Synonym n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163
PubChem CID 12742
IUPAC Name 1-bromononane
SMILES CCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 201°C
Quantity 25 mL
Formula Weight 207.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromononane
EINECSNumber (2)-0067
TSCA Yes