missing translation for 'onlineSavingsMsg'
Learn More

1-Bromononane, 99%

Catalog No. AAA1057714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1057714 25 g
AAA1057722 100 g
AAA1057736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1057714 Supplier Thermo Scientific Chemicals Supplier No. A1057714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 693-58-3 | C9H19Br | 207.155 g/mol

1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

Solubility
Insoluble in water. Soluble in ethyl ether and benzene.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 693-58-3
Molecular Formula C9H19Br
Molecular Weight (g/mol) 207.155
MDL Number MFCD00000278
InChI Key AYMUQTNXKPEMLM-UHFFFAOYSA-N
Synonym n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163
PubChem CID 12742
IUPAC Name 1-bromononane
SMILES CCCCCCCCCBr

Specifications

Density 1.086
Boiling Point 200°C to 201°C
Flash Point 90°C (194°F)
Refractive Index 1.455
Quantity 25 g
Beilstein 1733632
Solubility Information Insoluble in water. Soluble in ethyl ether and benzene.
Formula Weight 207.16
Percent Purity 99%
Chemical Name or Material 1-Bromononane
Hazard Category H227-H315-H319-H332-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-755-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only