missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)naphthalene 98.0+%, TCI America™

Catalog No. p-7123619
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45635G 5 g
B456325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45635G Supplier TCI America Supplier No. B45635G
Only null left

Chemical Identifiers

CAS 3163-27-7
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.097
MDL Number MFCD00010804
InChI Key RZJGKPNCYQZFGR-UHFFFAOYSA-N
Synonym 1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389
PubChem CID 137844
IUPAC Name 1-(bromomethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CBr

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 118°C
Quantity 5 g
UN Number 3261
Formula Weight 221.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Bromomethyl)naphthalene
TSCA No
Recommended Storage Refrigerator