missing translation for 'onlineSavingsMsg'
Learn More

1-(bromomethyl)-2-phenoxybenzene, 90%, Thermo Scientific™

Catalog No. p-4510711
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC49908DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC49908DA Supplier Thermo Scientific Chemicals Supplier No. CC49908DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82657-72-5
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
InChI Key YQRIQBOWLXRKKG-UHFFFAOYSA-N
Synonym 1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
PubChem CID 22675469
IUPAC Name 1-(bromomethyl)-2-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CBr

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-2-phenoxybenzene