missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoisoquinoline 98.0+%, TCI America™
$359.76 - $1077.43
Chemical Identifiers
| CAS | 1532-71-4 |
|---|---|
| Molecular Formula | C9H6BrN |
| Molecular Weight (g/mol) | 208.058 |
| MDL Number | MFCD00234478 |
| InChI Key | YWWZASFPWWPUBN-UHFFFAOYSA-N |
| Synonym | isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888 |
| PubChem CID | 640963 |
| IUPAC Name | 1-bromoisoquinoline |
| SMILES | C1=CC=C2C(=C1)C=CN=C2Br |
Chemical Identifiers
| 1532-71-4 | |
| 208.058 | |
| YWWZASFPWWPUBN-UHFFFAOYSA-N | |
| 640963 | |
| C1=CC=C2C(=C1)C=CN=C2Br |
| C9H6BrN | |
| MFCD00234478 | |
| isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888 | |
| 1-bromoisoquinoline |
Specifications
| 1532-71-4 | |
| White-Yellow | |
| MFCD00234478 | |
| isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888 | |
| C1=CC=C2C(=C1)C=CN=C2Br | |
| 208.058 | |
| 208.06 | |
| Crystalline Powder |
| 44°C | |
| C9H6BrN | |
| 1 g | |
| YWWZASFPWWPUBN-UHFFFAOYSA-N | |
| 1-bromoisoquinoline | |
| 640963 | |
| ≥98.0% (T) | |
| 1-Bromoisoquinoline |
Safety and Handling
TSCA : No