accessibility menu, dialog, popup

UserName

BrN

CAS RN 1532-71-4

IUPAC Name: 1-bromoisoquinoline
Synonyms: 1-bromoisoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: YWWZASFPWWPUBN-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)