missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoisoquinoline, 95+%, Thermo Scientific™

Chemical Identifiers

CAS 1532-71-4
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.05
MDL Number MFCD00234478
InChI Key YWWZASFPWWPUBN-UHFFFAOYSA-N
Synonym isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888
PubChem CID 640963
IUPAC Name 1-bromoisoquinoline
SMILES C1=CC=C2C(=C1)C=CN=C2Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC392590010
SDS View Documents
Thermo Scientific Chemicals
392590010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1532-71-4
208.05
YWWZASFPWWPUBN-UHFFFAOYSA-N
640963
C1=CC=C2C(=C1)C=CN=C2Br
C9H6BrN
MFCD00234478
isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888
1-bromoisoquinoline

Specifications

1532-71-4
100.0
95% min. (HPLC)
C9H6BrN
1g
YWWZASFPWWPUBN-UHFFFAOYSA-N
1-bromoisoquinoline
640963
95+%
95.0
Authentic
Glass bottle
MFCD00234478
isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888
C1=CC=C2C(=C1)C=CN=C2Br
208.05
208.05
1-Bromoisoquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only