missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexane 98.0+%, TCI America™

Catalog No. B0600500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0600500G 500 g
B060025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0600500G Supplier TCI America Supplier No. B0600500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-25-1
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.07
MDL Number MFCD00000271
InChI Key MNDIARAMWBIKFW-UHFFFAOYSA-N
Synonym hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
PubChem CID 8101
IUPAC Name 1-bromohexane
SMILES CCCCCCBr

Specifications

Melting Point -85°C
Color Yellow
Boiling Point 155°C
Quantity 500 g
UN Number 1993
Formula Weight 165.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromohexane
EINECSNumber (2)-0067
RTECSNumber MO0925000
TSCA Yes