missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexane, 99+%

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

Supplier:  Thermo Scientific Chemicals 107040025

 View more versions of this product

Catalog No. AC107040025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromohexane
0.1% max. (as HBr)
100.0
99+%
C6H13Br
CH3(CH2)5Br
2.5 L
MNDIARAMWBIKFW-UHFFFAOYSA-N
1-bromohexane
8101
≥99.0%
111-25-1
99.0
Authentic
Glass bottle
1.4465 to 1.4485 (20°C, 589nm)
MFCD00000271
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
CCCCCCBr
165.07
165.07
Liquid

Chemical Identifiers

111-25-1
165.07
MNDIARAMWBIKFW-UHFFFAOYSA-N
8101
CCCCCCBr
C6H13Br
MFCD00000271
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
1-bromohexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only