Learn More
1-Bromohexane, 99%
CAS: 111-25-1 | C6H13Br | 165.07 g/mol
Supplier: Thermo Scientific Chemicals A151660E
Description
1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.
Solubility
Immiscible with water.
Notes
Keep away from heat and sources of ignition.
Specifications
| 1-Bromohexane | |
| -85°C | |
| 156°C | |
| Pleasant | |
| 1.4475 | |
| 2500 g | |
| 1731290 | |
| Immiscible with water. | |
| CCCCCCBr | |
| 165.07 | |
| 165.08 |
| 111-25-1 | |
| 1.176 | |
| 50°C (122°F) | |
| C6H13Br | |
| MFCD00000271 | |
| UN1993 | |
| hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga | |
| MNDIARAMWBIKFW-UHFFFAOYSA-N | |
| 1-bromohexane | |
| 8101 | |
| 99% |
Chemical Identifiers
| 111-25-1 | |
| 165.07 | |
| MNDIARAMWBIKFW-UHFFFAOYSA-N | |
| 8101 | |
| CCCCCCBr |
| C6H13Br | |
| MFCD00000271 | |
| hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga | |
| 1-bromohexane |
Safety and Handling
GHS H Statement
H226-H315
Flammable liquid and vapour.
Causes skin irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 203-850-2
RTECSNumber : MO0925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only