missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexane, 99%

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

Supplier:  Thermo Scientific Chemicals A151660C

 View more versions of this product

Catalog No. AAA151660C

Only null left
Explore available promotions

Description

Description

1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

Solubility
Immiscible with water.

Notes
Keep away from heat and sources of ignition.
Specifications

Specifications

1-Bromohexane
-85°C
156°C
Pleasant
1.4475
10,000 g
1731290
Immiscible with water.
CCCCCCBr
165.07
165.08
111-25-1
1.176
50°C (122°F)
C6H13Br
MFCD00000271
UN1993
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
MNDIARAMWBIKFW-UHFFFAOYSA-N
1-bromohexane
8101
99%

Chemical Identifiers

111-25-1
165.07
MNDIARAMWBIKFW-UHFFFAOYSA-N
8101
CCCCCCBr
C6H13Br
MFCD00000271
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
1-bromohexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315
Flammable liquid and vapour.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 203-850-2

RTECSNumber : MO0925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only