missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromohexadecane 96.0+%, TCI America™
$29.83 - $130.48
Chemical Identifiers
| CAS | 112-82-3 |
|---|---|
| Molecular Formula | C16H33Br |
| Molecular Weight (g/mol) | 305.34 |
| MDL Number | MFCD00000230 |
| InChI Key | HNTGIJLWHDPAFN-UHFFFAOYSA-N |
| Synonym | hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam |
| PubChem CID | 8213 |
| IUPAC Name | 1-bromohexadecane |
| SMILES | CCCCCCCCCCCCCCCCBr |
Chemical Identifiers
| 112-82-3 | |
| 305.34 | |
| HNTGIJLWHDPAFN-UHFFFAOYSA-N | |
| 8213 | |
| CCCCCCCCCCCCCCCCBr |
| C16H33Br | |
| MFCD00000230 | |
| hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam | |
| 1-bromohexadecane |
Specifications
| 112-82-3 | |
| 197°C | |
| MFCD00000230 | |
| hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam | |
| CCCCCCCCCCCCCCCCBr | |
| 305.34 | |
| 305.34 | |
| Liquid |
| Yellow | |
| C16H33Br | |
| 25 g | |
| HNTGIJLWHDPAFN-UHFFFAOYSA-N | |
| 1-bromohexadecane | |
| 8213 | |
| ≥96.0% (GC) | |
| 1-Bromohexadecane |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes