missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexadecane 96.0+%, TCI America™

Catalog No. B059925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059925G 25 g
B0599100G 100 g
B0599500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B059925G Supplier TCI America Supplier No. B059925G

Chemical Identifiers

CAS 112-82-3
Molecular Formula C16H33Br
Molecular Weight (g/mol) 305.34
MDL Number MFCD00000230
InChI Key HNTGIJLWHDPAFN-UHFFFAOYSA-N
Synonym hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
PubChem CID 8213
IUPAC Name 1-bromohexadecane
SMILES CCCCCCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 197°C
Quantity 25 g
Formula Weight 305.34
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromohexadecane
EINECSNumber (2)-0067
TSCA Yes