missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexadecane, 97%

Catalog No. p-4509965
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215432500 250 mL Glass bottle
AC215430010 1 L Glass bottle
AC215430025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC215432500 Supplier Thermo Scientific Chemicals Supplier No. 215432500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-82-3 | C16H33Br | 305.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-82-3
Molecular Formula C16H33Br
Molecular Weight (g/mol) 305.34
MDL Number MFCD00000230
InChI Key HNTGIJLWHDPAFN-UHFFFAOYSA-N
Synonym hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
PubChem CID 8213
IUPAC Name 1-bromohexadecane
SMILES CCCCCCCCCCCCCCCCBr

Specifications

Melting Point 17.3°C
Color Amber to Black-Brown
Density 0.9990g/mL
Boiling Point 336.0°C
Flash Point 177°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4609 to 1.4629
Linear Formula CH3(CH2)15Br
Quantity 250 mL
Beilstein 01,172
Specific Gravity 0.999
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 305.33
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 1-Bromohexadecane
Show More Show Less
EINECSNumber 204-008-7

RUO – Research Use Only