missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoheptane 98.0+%, TCI America™
Supplier: TCI America B0598500G
Specifications
| 1-Bromoheptane | |
| -58°C | |
| 180°C | |
| MFCD00000273 | |
| 1993 | |
| LSXKDWGTSHCFPP-UHFFFAOYSA-N | |
| 1-bromoheptane | |
| 12369 | |
| ≥98.0% (GC) |
| 629-04-9 | |
| Yellow | |
| C7H15Br | |
| 500 g | |
| heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane | |
| CCCCCCCBr | |
| 179.10 | |
| 179.10 | |
| Liquid |
Chemical Identifiers
| 629-04-9 | |
| 179.10 | |
| LSXKDWGTSHCFPP-UHFFFAOYSA-N | |
| 12369 | |
| CCCCCCCBr |
| C7H15Br | |
| MFCD00000273 | |
| heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane | |
| 1-bromoheptane |
Safety and Handling
EINECSNumber : (2)-0067
RTECSNumber : MI8100000
TSCA : Yes