missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoheptane, 98+%

Catalog No. AAA1403130
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1403130 250 g
AAA140310B 1000 g
AAA140310I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1403130 Supplier Thermo Scientific Chemicals Supplier No. A1403130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-04-9 | C7H15Br | 179.10 g/mol

1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Immiscible with water.

Notes
Flammable and incompatible with oxidizing agents, strong bases.

Chemical Identifiers

CAS 629-04-9
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.10
MDL Number MFCD00000273
InChI Key LSXKDWGTSHCFPP-UHFFFAOYSA-N
Synonym heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane
PubChem CID 12369
IUPAC Name 1-bromoheptane
SMILES CCCCCCCBr

Specifications

Melting Point -58°C
Density 1.139
Boiling Point 179°C to 180°C
Flash Point 60°C (140°F)
Odor Recognizable
Refractive Index 1.4499
Quantity 250 g
UN Number UN1993
Beilstein 1697160
Solubility Information Immiscible with water.
Formula Weight 179.11
Percent Purity ≥98%
Chemical Name or Material 1-Bromoheptane
Show More Show Less
Hazard Category H226-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 211-068-8
RTECSNumber MI8100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only