missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoheptadecane 98.0+%, TCI America™

Catalog No. B09375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09375G 5 g
B093725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09375G Supplier TCI America Supplier No. B09375G
Only null left

Chemical Identifiers

CAS 3508-00-7
Molecular Formula C17H35Br
Molecular Weight (g/mol) 319.371
MDL Number MFCD00017891
InChI Key HHSDZLLPIXMEIU-UHFFFAOYSA-N
Synonym Heptadecyl Bromide
PubChem CID 19022
IUPAC Name 1-bromoheptadecane
SMILES CCCCCCCCCCCCCCCCCBr

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 319.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromoheptadecane
EINECSNumber (2)-0067
TSCA Yes