missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Bromoethyl)benzene 95.0+%, TCI America™
$55.84 - $159.79
Chemical Identifiers
| CAS | 585-71-7 |
|---|---|
| Molecular Formula | C8H9Br |
| Molecular Weight (g/mol) | 185.064 |
| MDL Number | MFCD00000139 |
| InChI Key | CRRUGYDDEMGVDY-UHFFFAOYSA-N |
| Synonym | 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene |
| PubChem CID | 11454 |
| IUPAC Name | 1-bromoethylbenzene |
| SMILES | CC(C1=CC=CC=C1)Br |
Chemical Identifiers
| 585-71-7 | |
| 185.064 | |
| CRRUGYDDEMGVDY-UHFFFAOYSA-N | |
| 11454 | |
| CC(C1=CC=CC=C1)Br |
| C8H9Br | |
| MFCD00000139 | |
| 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene | |
| 1-bromoethylbenzene |
Specifications
| 585-71-7 | |
| 200°C | |
| MFCD00000139 | |
| 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene | |
| CC(C1=CC=CC=C1)Br | |
| 185.064 | |
| 185.06 | |
| Liquid |
| Yellow | |
| C8H9Br | |
| 25 g | |
| CRRUGYDDEMGVDY-UHFFFAOYSA-N | |
| 1-bromoethylbenzene | |
| 11454 | |
| ≥95.0% (GC) | |
| (1-Bromoethyl)benzene |
Safety and Handling
EINECSNumber : (3)-0035
TSCA : Yes
Recommended Storage : Refrigerator