missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene 95.0+%, TCI America™

Catalog No. B059225G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059225G 25 g
B0592100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B059225G Supplier TCI America Supplier No. B059225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 g
Formula Weight 185.06
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (1-Bromoethyl)benzene
EINECSNumber (3)-0035
TSCA Yes
Recommended Storage Refrigerator