missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene, 97%

Catalog No. p-7023367
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1683914 25 g
AAA1683922 100 g
AAA1683936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1683914 Supplier Thermo Scientific Chemicals Supplier No. A1683914

CAS: 585-71-7 | C8H9Br | 185.064 g/mol

(1-Bromoethyl)benzene has been employed in controlled radical polymerization of styrene, in asymmetric esterification of benzoic acid in the presence of a chiral cyclic guanidine and as initiator in the synthesis of bromine terminated polyp-methoxystyrene and polystyrene via atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Bromoethyl)benzene has been employed in controlled radical polymerization of styrene, in asymmetric esterification of benzoic acid in the presence of a chiral cyclic guanidine and as initiator in the synthesis of bromine terminated polyp-methoxystyrene and polystyrene via atom transfer radical polymerization.

Solubility
Soluble in alcohol, ether, and benzene.

Notes
Moisture Sensitive. Do not store together with strongly basic and oxidizing materials.

Chemical Identifiers

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br

Specifications

Melting Point -65°C
Density 1.356
Boiling Point 200°C
Flash Point 81°C (177°F)
Refractive Index 1.56
Quantity 25 g
Beilstein 507210
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohol,ether,and benzene.
Formula Weight 185.06
Percent Purity 97%
Chemical Name or Material (1-Bromoethyl)benzene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 209-560-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only