missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene, 97%

CAS: 585-71-7 | C8H9Br | 185.06 g/mol

$374.13 - $374.13

Chemical Identifiers

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218391000
SDS View Documents
Thermo Scientific Chemicals
218391000
100 g Glass bottle
Each for $374.13
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

585-71-7
185.06
CRRUGYDDEMGVDY-UHFFFAOYSA-N
11454
CC(C1=CC=CC=C1)Br
C8H9Br
MFCD00000139
1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
1-bromoethylbenzene

Specifications

585-71-7
100.0
1.3500g/mL
81°C
96% min. (GC)
C8H9Br
C6H5CH(CH3)Br
100 g
1.35
CRRUGYDDEMGVDY-UHFFFAOYSA-N
1-bromoethylbenzene
11454
97%
(1-Bromoethyl)benzene
96.0
Brown to Yellow
94.0°C (16.0 mmHg)
Authentic
Glass bottle
1.5593 to 1.5613
MFCD00000139
05, 355
1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
CC(C1=CC=CC=C1)Br
185.06
185.06
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 209-560-2

SDS
Documents

Documents

RUO – Research Use Only