missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene, 97%

Catalog No. AC218390250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218390250 25 g Glass bottle
AC218391000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218390250 Supplier Thermo Scientific Chemicals Supplier No. 218390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 585-71-7 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.3500g/mL
Boiling Point 94.0°C (16.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5593 to 1.5613
Linear Formula C6H5CH(CH3)Br
Quantity 25 g
Beilstein 05, 355
Specific Gravity 1.35
Formula Weight 185.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (1-Bromoethyl)benzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 209-560-2

RUO – Research Use Only