missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoeicosane 95.0+%, TCI America™

Catalog No. B039125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B039125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B039125G Supplier TCI America Supplier No. B039125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4276-49-7
Molecular Formula C20H41Br
Molecular Weight (g/mol) 361.45
MDL Number MFCD00013542
InChI Key CZASMUMJSKOHFJ-UHFFFAOYSA-N
Synonym Eicosyl Bromide
PubChem CID 20271
IUPAC Name 1-bromoicosane
SMILES CCCCCCCCCCCCCCCCCCCCBr

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 386°C
Quantity 25 g
Formula Weight 361.45
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromoeicosane
EINECSNumber (2)-0067
TSCA Yes
Recommended Storage Refrigerator