missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane 98.0+%, TCI America™

Catalog No. p-7118544
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
B058725ML 25 mL
B0587500ML 500 mL
2 options

Catalog No. B058725ML

Supplier: TCI America B058725ML

Only null left

Chemical Identifiers

CAS 143-15-7
Molecular Formula C12H25Br
Molecular Weight (g/mol) 249.236
MDL Number MFCD00000225
InChI Key PBLNBZIONSLZBU-UHFFFAOYSA-N
Synonym dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
PubChem CID 8919
IUPAC Name 1-bromododecane
SMILES CCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 mL
Formula Weight 249.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromododecane
EINECSNumber (2)-0067
TSCA Yes