missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane 98.0+%, TCI America™

Catalog No. B0587500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0587500ML 500 mL
B058725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0587500ML Supplier TCI America Supplier No. B0587500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143-15-7
Molecular Formula C12H25Br
Molecular Weight (g/mol) 249.236
MDL Number MFCD00000225
InChI Key PBLNBZIONSLZBU-UHFFFAOYSA-N
Synonym dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
PubChem CID 8919
IUPAC Name 1-bromododecane
SMILES CCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 140°C
Quantity 500 mL
Formula Weight 249.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromododecane
EINECSNumber (2)-0067
TSCA Yes