Learn More
1-Bromododecane, 98%
CAS: 143-15-7 | C12H25Br | 249.236 g/mol
Supplier: Thermo Scientific Chemicals A106000I
Description
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 1-Bromododecane | |
| -10°C | |
| 276°C | |
| Weak | |
| 1.458 | |
| MFCD00000225 | |
| 506159 | |
| dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d | |
| PBLNBZIONSLZBU-UHFFFAOYSA-N | |
| 1-bromododecane | |
| 8919 | |
| 98% |
| 143-15-7 | |
| 1.04 | |
| 146°C (294°F) | |
| C12H25Br | |
| CH3(CH2)11Br | |
| 5000 g | |
| 14,5389 | |
| Insoluble in water. | |
| CCCCCCCCCCCCBr | |
| 249.236 | |
| 249.24 |
Chemical Identifiers
| 143-15-7 | |
| 249.236 | |
| PBLNBZIONSLZBU-UHFFFAOYSA-N | |
| 8919 | |
| CCCCCCCCCCCCBr |
| C12H25Br | |
| MFCD00000225 | |
| dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d | |
| 1-bromododecane |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-587-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only