missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane, 98%

Catalog No. AAA1060030
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1060030 250 g
AAA106000B 1000 g
AAA106000I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1060030 Supplier Thermo Scientific Chemicals Supplier No. A1060030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 143-15-7 | C12H25Br | 249.236 g/mol

1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 143-15-7
Molecular Formula C12H25Br
Molecular Weight (g/mol) 249.236
MDL Number MFCD00000225
InChI Key PBLNBZIONSLZBU-UHFFFAOYSA-N
Synonym dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
PubChem CID 8919
IUPAC Name 1-bromododecane
SMILES CCCCCCCCCCCCBr

Specifications

Melting Point -10°C
Density 1.04
Boiling Point 276°C
Flash Point 146°C (294°F)
Odor Weak
Refractive Index 1.458
Linear Formula CH3(CH2)11Br
Quantity 250 g
Beilstein 506159
Merck Index 14,5389
Solubility Information Insoluble in water.
Formula Weight 249.24
Percent Purity 98%
Chemical Name or Material 1-Bromododecane
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-587-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only