missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromodocosane 98.0+%, TCI America™
$156.59 - $976.76
Chemical Identifiers
| CAS | 6938-66-5 |
|---|---|
| Molecular Formula | C22H45Br |
| Molecular Weight (g/mol) | 389.506 |
| MDL Number | MFCD00013543 |
| InChI Key | QYOXLKAKUAASNA-UHFFFAOYSA-N |
| Synonym | Docosyl Bromide |
| PubChem CID | 81355 |
| IUPAC Name | 1-bromodocosane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCBr |
Chemical Identifiers
| 6938-66-5 | |
| 389.506 | |
| QYOXLKAKUAASNA-UHFFFAOYSA-N | |
| 81355 | |
| CCCCCCCCCCCCCCCCCCCCCCBr |
| C22H45Br | |
| MFCD00013543 | |
| Docosyl Bromide | |
| 1-bromodocosane |
Specifications
| 6938-66-5 | |
| White-Yellow | |
| MFCD00013543 | |
| Docosyl Bromide | |
| CCCCCCCCCCCCCCCCCCCCCCBr | |
| 389.506 | |
| 389.51 | |
| Crystalline Powder |
| 44°C | |
| C22H45Br | |
| 25 g | |
| QYOXLKAKUAASNA-UHFFFAOYSA-N | |
| 1-bromodocosane | |
| 81355 | |
| ≥98.0% (GC) | |
| 1-Bromodocosane |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes
Recommended Storage : Refrigerator