missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromodocosane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 44°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 389.51 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Bromodocosane |
Safety and Handling
| EINECSNumber | (2)-0067 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |